ENA183042205 Enamine

1-(4-methylpentan-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD09740703
  • Cas Number : 1052566-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-38936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-38936
Availability details for
Quantity
  • Availability
  • Add to Cart