ENA183042207 Enamine

1-(pyridin-2-yl)piperidin-4-one

  • MDL number MFCD09945314
  • Cas Number : 264608-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39460
Availability details for
Quantity
  • Availability
  • Add to Cart