ENA183042210 Enamine

2-(1H-1,3-benzodiazol-1-yl)pyridine-3-carboxylic acid

  • MDL number MFCD09742241
  • Cas Number : 954569-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39075
Availability details for
Quantity
  • Availability
  • Add to Cart