ENA183042212 Enamine

3-[(1,3-benzothiazol-2-ylthio)methyl]benzoic acid

  • MDL number MFCD06628914
  • Cas Number : 832688-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39466

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39466
Availability details for
Quantity
  • Availability
  • Add to Cart