ENA183042234 Enamine

5-(piperazin-1-ylsulfonyl)-1,3-dihydro-2H-benzimidazol-2-one

  • MDL number MFCD08271816
  • Cas Number : 900641-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39503

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39503
Availability details for
Quantity
  • Availability
  • Add to Cart