ENA183042240 Enamine

4-methyl-5-(morpholin-4-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD09041978
  • Cas Number : 926269-71-8
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39220
Availability details for
Quantity
  • Availability
  • Add to Cart