ENA183042243 Enamine

4-{[(pyridin-3-yl)methyl]sulfanyl}benzoic acid

  • MDL number MFCD09737036
  • Cas Number : 954269-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39514
Availability details for
Quantity
  • Availability
  • Add to Cart