ENA183042248 Enamine

3-(aminomethyl)-N,N-dimethylpyridin-2-amine

  • MDL number MFCD09731450
  • Cas Number : 354824-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39521
Availability details for
Quantity
  • Availability
  • Add to Cart