ENA183042258 Enamine

1-[(oxolan-2-yl)methyl]piperidin-4-amine

  • MDL number MFCD09047072
  • Cas Number : 108282-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39532
Availability details for
Quantity
  • Availability
  • Add to Cart