ENA183042273 Enamine

4-methyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD00030418
  • Cas Number : 64276-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39551
Availability details for
Quantity
  • Availability
  • Add to Cart