ENA183042278 Enamine

N-1,3-benzodioxol-5-yl-2,2,2-trichloroacetamide

  • MDL number MFCD01346393
  • Cas Number : 549543-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39241

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39241
Availability details for
Quantity
  • Availability
  • Add to Cart