ENA183042295 Enamine

1-(benzylsulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD09049310
  • Cas Number : 891392-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39584

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39584
Availability details for
Quantity
  • Availability
  • Add to Cart