ENA183042301 Enamine

4-amino-N-(2-methoxyethyl)benzenesulfonamide

  • MDL number MFCD09703303
  • Cas Number : 328062-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83...85
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39591

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39591
Availability details for
Quantity
  • Availability
  • Add to Cart