ENA183042302 Enamine

5,6-dimethyl-1-(4-methylbenzyl)-1H-benzimidazole

  • MDL number MFCD04217361
  • Cas Number : 141211-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39592

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39592
Availability details for
Quantity
  • Availability
  • Add to Cart