ENA183042303 Enamine

1-(4-tert-butylbenzyl)-5,6-dimethyl-1H-benzimidazole

  • MDL number MFCD06643536
  • Cas Number : 871666-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39593

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39593
Availability details for
Quantity
  • Availability
  • Add to Cart