ENA183042306 Enamine

4-[(2,5-dichlorophenoxy)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD11505400
  • Cas Number : 1082303-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39263
Availability details for
Quantity
  • Availability
  • Add to Cart