ENA183042307 Enamine

4-(2-oxo-1,3-oxazolidin-3-yl)benzoic acid

  • MDL number MFCD11167850
  • Cas Number : 208708-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39599
Availability details for
Quantity
  • Availability
  • Add to Cart