ENA183042312 Enamine

3-amino-N-(1,1-dioxo-1lambda6-thiolan-3-yl)-4-methoxybenzamide

  • MDL number MFCD09811858
  • Cas Number : 1016741-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39609
Availability details for
Quantity
  • Availability
  • Add to Cart