ENA183042315 Enamine

1-(4-chlorobenzyl)-5,6-dimethyl-1H-benzimidazole

  • MDL number MFCD01304890
  • Cas Number : 141472-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39612

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39612
Availability details for
Quantity
  • Availability
  • Add to Cart