ENA183042320 Enamine

1-(4-fluorobenzyl)-5,6-dimethyl-1H-benzimidazole

  • MDL number MFCD04215631
  • Cas Number : 637325-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39620

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39620
Availability details for
Quantity
  • Availability
  • Add to Cart