ENA183042324 Enamine

1-iodo-2-methyl-3-nitrobenzene

  • MDL number MFCD08272202
  • Cas Number : 41252-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39630
Availability details for
Quantity
  • Availability
  • Add to Cart