ENA183042325 Enamine

2-sulfanylpyridine-3-carboxamide

  • MDL number MFCD01570797
  • Cas Number : 50596-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-39631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39631
Availability details for
Quantity
  • Availability
  • Add to Cart