ENA183042328 Enamine

N-(3-fluorophenyl)-3-oxobutanamide

  • MDL number MFCD00232365
  • Cas Number : 52173-35-0
  • Molecular Weight : 195.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39637
Availability details for
Quantity
  • Availability
  • Add to Cart