ENA183042330 Enamine

3-amino-4-methoxy-N-(5-methyl-1,3-thiazol-2-yl)benzamide

  • MDL number MFCD09730805
  • Cas Number : 953744-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39639
Availability details for
Quantity
  • Availability
  • Add to Cart