ENA183042335 Enamine

8-oxatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),3,5,10,12-hexaen-5-amine

  • MDL number MFCD00092339
  • Cas Number : 4106-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-93
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39645

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P312,P302+P352,P304+P340,P304+P341,P312,P330,P342+P311,P363,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-39645
pictograms GHS07,GHS08
hstatements H302,H312,H332,H334
Availability details for
Quantity
  • Availability
  • Add to Cart