ENA183042343 Enamine

amino(thien-2-yl)acetonitrile hydrochloride

  • MDL number MFCD11505497
  • Cas Number : 93554-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39656
Availability details for
Quantity
  • Availability
  • Add to Cart