ENA183042344 Enamine

4-[(pyridin-3-yl)methanesulfonyl]benzoic acid

  • MDL number MFCD09928320
  • Cas Number : 1016722-46-5
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39657
Availability details for
Quantity
  • Availability
  • Add to Cart