ENA183042347 Enamine

1-aminocyclobutanecarbonitrile hydrochloride

  • MDL number MFCD11505498
  • Cas Number : 845821-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39660
Availability details for
Quantity
  • Availability
  • Add to Cart