ENA183042350 Enamine

2-(4-cyano-2-methoxyphenoxy)-N,N-dimethylacetamide

  • MDL number MFCD07901184
  • Cas Number : 930937-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39663
Availability details for
Quantity
  • Availability
  • Add to Cart