ENA183042352 Enamine

4-(aminomethyl)-N-methylbenzene-1-sulfonamide

  • MDL number MFCD08337810
  • Cas Number : 101252-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39669
Availability details for
Quantity
  • Availability
  • Add to Cart