ENA183042358 Enamine

1-(2-methoxybenzyl)-5,6-dimethyl-1H-benzimidazole

  • MDL number MFCD08163114
  • Cas Number : 890634-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121...123
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39678

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39678
Availability details for
Quantity
  • Availability
  • Add to Cart