ENA183042359 Enamine

1-(2,4-difluorophenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD08444739
  • Cas Number : 125126-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39679

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39679
Availability details for
Quantity
  • Availability
  • Add to Cart