ENA183042364 Enamine

N-[3-(1-aminoethyl)phenyl]benzamide

  • MDL number MFCD09728810
  • Cas Number : 953725-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39692
Availability details for
Quantity
  • Availability
  • Add to Cart