ENA183042365 Enamine

1-cyclohexanecarbonylpiperidin-4-amine

  • MDL number MFCD09044156
  • Cas Number : 565453-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39693
Availability details for
Quantity
  • Availability
  • Add to Cart