ENA183042367 Enamine

1-(4-chlorophenyl)-6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD09047271
  • Cas Number : 926232-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39695

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39695
Availability details for
Quantity
  • Availability
  • Add to Cart