ENA183042371 Enamine

4-amino-N,3-dimethylbenzamide

  • MDL number MFCD09043972
  • Cas Number : 926263-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39699

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39699
Availability details for
Quantity
  • Availability
  • Add to Cart