ENA183042374 Enamine

[5-(1,3-benzothiazol-2-yl)-2-furyl]methylamine

  • MDL number MFCD08271855
  • Cas Number : 885527-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39703

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-39703
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart