ENA183042381 Enamine

1-[4-(aminomethyl)phenyl]-3-(propan-2-yl)urea

  • MDL number MFCD11167472
  • Cas Number : 1038238-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39714
Availability details for
Quantity
  • Availability
  • Add to Cart