ENA183042402 Enamine

4-phenylazetidin-2-one

  • MDL number MFCD02179052
  • Cas Number : 5661-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39746

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-39746
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart