ENA183042407 Enamine

2,2,2-trifluoroethyl chloridocarbonate

  • MDL number MFCD00800932
  • Cas Number : 27746-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31105

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-31105
pictograms GHS02,GHS05,GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart