ENA183042412 Enamine

2-methyl-3-(methylamino)propanenitrile

  • MDL number MFCD11505518
  • Cas Number : 50840-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39761
Availability details for
Quantity
  • Availability
  • Add to Cart