ENA183042423 Enamine

1-(2-amino-5-chlorobenzoyl)piperidine-3-carboxamide

  • MDL number MFCD09733338
  • Cas Number : 953905-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39775
Availability details for
Quantity
  • Availability
  • Add to Cart