ENA183042432 Enamine

1-methyl-4-{methyl[(pyridin-3-yl)methyl]sulfamoyl}-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09053130
  • Cas Number : 937629-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39791
Availability details for
Quantity
  • Availability
  • Add to Cart