ENA183042439 Enamine

1-(5-bromofuran-2-carbonyl)piperazine

  • MDL number MFCD08444697
  • Cas Number : 66204-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39801
Availability details for
Quantity
  • Availability
  • Add to Cart