ENA183042444 Enamine

3,3-dimethyl-1-phenylbutan-1-one

  • MDL number MFCD03841073
  • Cas Number : 31366-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39809
Availability details for
Quantity
  • Availability
  • Add to Cart