ENA183042449 Enamine

3-(3-amino-4-methylphenyl)-1-phenylurea

  • MDL number MFCD09951030
  • Cas Number : 691897-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39818
Availability details for
Quantity
  • Availability
  • Add to Cart