ENA183042453 Enamine

3-(propan-2-yl)-3-azabicyclo[3.3.1]nonan-9-ol

  • MDL number MFCD13806378
  • Cas Number : 1177337-43-7
  • Molecular Weight : 183.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39823
Availability details for
Quantity
  • Availability
  • Add to Cart