ENA183042457 Enamine

4-phenyl-5-(propan-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD09733229
  • Cas Number : 953732-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39834
Availability details for
Quantity
  • Availability
  • Add to Cart