ENA183042461 Enamine

N-[(piperidin-2-yl)methyl]methanesulfonamide

  • MDL number MFCD09814145
  • Cas Number : 1016509-49-1
  • Molecular Weight : 192.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31772
Availability details for
Quantity
  • Availability
  • Add to Cart