ENA183042476 Enamine

2-[(3-bromobenzyl)thio]nicotinic acid

  • MDL number MFCD09732783
  • Cas Number : 450385-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39878

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39878
Availability details for
Quantity
  • Availability
  • Add to Cart